Anti-MRC1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Supplier: Bioss

BS-4727R-A750
76077-724EA 1190.99 CAD
76077-724
Anti-MRC1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antibodies
Mediates the endocytosis of glycoproteins by macrophages. Binds both sulfated and non-sulfated polysaccharide chains. Acts as phagocytic receptor for bacteria, fungi and other pathogens.

Type: Primary
Antigen: MRC1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 201-300/1456
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat; Dog
Order Now

SPECIFICATIONS

Environmentally Preferable
Antigen name MRC1
Antigen synonyms CD206|bA541I19.1|C-type lectin domain family 13 member D-like|Macrophage mannose receptor 1|CLEC13D|MRC1|MRC1L1|MMR|C-type lectin domain family 13 member D|CLEC13DL|Macrophage mannose receptor 1-like protein 1
Antibody type Primary
Clonality Polyclonal
Conjugation Alexa Fluor® 750
Reactivity Human,Dog,Rat,Mouse
Host Rabbit
Gene ID 4360
Isotype IgG
Western blot Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Flow cytometry Yes
Modification Unmodified
Epitope 201-300/1456
Immunogen KLH conjugated synthetic peptide derived from human MRC1
Form Liquid
Molecular weight 160 kDa
Purification Purified by Protein A
Concentration 1 µg/µl
Storage buffer Aqueous buffered solution containing 0.01 M TBS (pH 7.4) with 1% BSA, 0.03% Proclin300 and 50% Glycerol
Storage temperature Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Shipping temperature 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR