LKV Agar, Reducible, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
A60
75846-312PK 59.96 CAD
75846-312
LKV Agar, Reducible, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
LKV (Laked Sheep Blood, Kanamycin, Vancomycin) Agar, reducible, for anaerobes.
  • Selective isolation of gram negative anaerobic bacteria
  • Laked Sheep Blood, Kanamycin and Vancomycin agar
  • Reducible, for anaerobes
(Brucella Agar with Laked Sheep Blood, Kanamycin, Vancomycin, Hemin and Vitamin K) For the selective isolation of gram negative anaerobic bacteria, especially Bacteroides spp.