TE (Tris EDTA), 1X, pH 8.0 (Nuclease Free)
Supplier: GROWCELLS
MRGF-4240 MRGF-4241 MRGF-4240-10L MRGF-4240-10Q MRGF-4240-20Q
75800-970CS 1375.07 CAD
75800-970 75800-972 75800-974 75800-976 75800-978 75800-980
TE (Tris EDTA), 1X, pH 8.0 (Nuclease Free)
Buffers Cloning Buffers
Nuclease free TE is used to solubilize DNA/RNA while providing protection from degradation
It provides great buffering capabilites for electrophoresis, and can be used for general storage of nucleic acids.
Order Now
SPECIFICATIONS
Environmentally Preferable