Human Recombinant IL1B (from E. coli)
Supplier: Prosci
90-463
75794-472EA 1639.38 CAD
75794-472
Human Recombinant IL1B (from E. coli)
Proteins and Peptides
IL-1beta is produced by activated macrophages. IL-1beta stimulates thymocyte proliferation by inducing IL-2 release, B-cell maturation and proliferation, and fibroblast growth factor activity. IL-1beta belongs to the IL-1 family of proteins that are involved in the inflammatory response. IL-1beta in most situations requires the inflammasome complex to be cleaved and secreted.
  • Fusion-Tag: His Tag
This recombinant proteins is for research use only.