Mouse Recombinant FGF21 (from HEK293 Cells)
Supplier: Prosci
90-323
75793-848EA 820.92 CAD
75793-848
Mouse Recombinant FGF21 (from HEK293 Cells)
Proteins and Peptides
Fibroblast growth factor 21 (FGF-21) is a member of the fibroblast growth factor family and has been identified as an important regulator of energy metabolism connecting nutrition, growth, reproduction and longevity. FGF-21 lacks the heparin-binding domain of most FGF proteins, allowing its secretion. FGF-21 is abundantly expressed in adipose tissues, liver and pancreas. In adipose tissue, FGF-21 promotes glucose uptake and oxidation and in liver it replenishes on fasting the tissues with fuel on low nutritional supply. It also induces lipolysis and ketogenesis. FGF-21 acts through FGF receptors (FGFR) associated with the auxiliary protein beta-Klotho. Recently, FGF-21 has been reported to interact directly with the brain circadian clock to coordinate activity and reproduction as part of the adaptation to fasting. Due to its multiple functions of normalizing glucose, lipid and energy homeostasis, FGF-21 represents an attractive novel therapy for type 2 diabetes mellitus and obesity.
  • Fusion-Tag: DDDDK Tag
This recombinant proteins is for research use only.