Anti-IDO1 Rabbit Polyclonal Antibody
Supplier: Prosci
36-228
75793-316EA 984.91 CAD
75793-316
Anti-IDO1 Rabbit Polyclonal Antibody
Antibodies
IDO catalyzes the first and rate-limiting step in the main pathway of human tryptophan catabolism, the kynurenine pathway. Proinflammatory mediators, such as endotoxin and IFN-gamma induce the expression of IDO in several tissues. IDO-dependent suppression of T-cell responses might function as natural immunoregulatory mechanism. Physiological IDO activity has been implicated in T-cell tolerance to tumors, dysfunctional selftolerance in non-obese diabetic (NOD) mice, and as a protective negative regulator in autoimmune disorders.
ELISA: (direct and indirect: 1:2,000-1:10,000). FlowMore Product Information