Human Recombinant Sialic Acid Synthase (from E. coli)
Supplier: Prosci
92-137
75790-868EA 1199.09 CAD
75790-868
Human Recombinant Sialic Acid Synthase (from E. coli)
Enzymes
Sialic Acid Synthase (NANS) is an enzyme that contains one AFP-like domain
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
NANS is ubiquitous and plays a role in the biosynthetic pathways of sialic acids. NANS produces N-acetylneuraminic acid (Neu5Ac) and 2-keto-3-deoxy-D-glycero-D-galacto-nononic acid (KDN). It also can use N-acetylmannosamine 6-phosphate and mannose 6-phosphate as substrates to generate phosphorylated forms of Neu5Ac and KDN, respectively.