Human Recombinant IGF2 mRNA-Binding 2 (from E. coli)
Supplier: Prosci
92-057
75790-708EA 1470.52 CAD
75790-708
Human Recombinant IGF2 mRNA-Binding 2 (from E. coli)
Proteins and Peptides
Insulin-Like Growth Factor 2 mRNA-Binding Protein 2 (IGFBP2) belongs to the RRM IMP/VICKZ family. IGFBP2 is a cytoplasmic protein and contains four KH domains and two RRM (RNA recognition motif) domains. IGF2BP2 binds to the 5'-UTR of the Insulin-Like Growth Factor 2 (IGF2) mRNA. This binding is isoform-specific. IGF2BP2 may regulate translation of target mRNAs. Genetic variation at the IGF2BP2 gene has been associated with type 2 diabetes (T2D) by genome-wide association studies and by replication analyses.
  • Fusion-Tag: C-6 His, N-T7 tag
This recombinant protein can be used for biological assays. For research use only.