Human Recombinant Zinc Finger 70 (fromE. coli)
Supplier: Prosci
92-033
75790-660EA 1470.52 CAD
75790-660
Human Recombinant Zinc Finger 70 (fromE. coli)
Proteins and Peptides
Zinc Finger Protein 70 (ZNF70) is a member of the krueppel C2H2-type zinc-finger protein family. ZFN70 is localized to the cell nucleus and contains eleven C2H2-type zinc fingers. ZFN70 has sequence-specific DNA binding transcription factor activity, and it may be involved in transcriptional regulation. In addition, ZFN70 has the DNA-binding and metal-binding activity, which can bind zinc ion.
  • Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.