Human Recombinant S100 calcium binding A8 (from E. coli)

Supplier: Prosci

91-636
75789-932EA 748.53 CAD
75789-932
Human Recombinant S100 calcium binding A8 (from E. coli)
Proteins and Peptides

Protein S100-A8 (also MRP8 and calgranulin A) is a calcium- and zinc-binding protein.It plays a prominent role in the regulation of inflammatory processes and immune response. S100A8 can induce neutrophil chemotaxis and adhesion. Its role as an oxidant scavenger has a protective role in preventing exaggerated tissue damage by scavenging oxidants. It can act as a potent amplifier of inflammation in autoimmunity as well as in cancer development and tumor spread.


  • Fusion-Tag: C-6 His tag


This recombinant protein can be used for biological assays. For research use only.

Order Now

SPECIFICATIONS

Protein/peptide name S100 calcium binding A8
Environmentally Preferable
Protein synonyms Mrp8|S100A8|Protein S100-A8|MRP-8|Leukocyte L1 complex light chain|Cystic fibrosis antigen|Calgranulin-A
Protein/peptide type Recombinant
Species Human
Source E. coli
Sequence Met1-Glu93
Endotoxin content <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
Purity > 95% as determined by reducing SDS-PAGE
Formulation Lyophilized from a 0.2 um filtered solution of 20mM HEPES,150mM NaCl, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
Molecular weight 11.7 kD
Storage conditions Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
Tag sequence C-6 His tag
Tested applications Bioassay

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR