Human Recombinant Carboxypeptidase B (from Cells)
Supplier: Prosci
91-411
75789-498EA 1525.33 CAD
75789-498
Human Recombinant Carboxypeptidase B (from Cells)
Enzymes
Carboxypeptidase B1 is an exopeptidase which specifically cleaves the C-terminal Arg and Lys residues with a preference for Arg
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Expressed mainly in pancreas, CPB1 is a useful serum marker for acute pancreatitis and pancreatic graft rejection.Human CPB1 consists of a signal peptide, a pro region, and a mature chain. The purified rhCPB1 corresponds to the pro form, which can be activated by trypsin, the only pancreatic protease capable of generating active enzyme from the zymogen in vitro.