3B Scientific® Birthing Simulators
Supplier: American 3B Scientific
1022878 1022879 1020337 1021567 1020336 1020338 1020339 1020340 1020341 1020342 1020343 1020344 1020345 1020346 1020347 1020348 1020608 1020367 1023327 1023328
470224-990EA 2569.7 CAD
470224-990 470224-992 470224-994 470311-936 470311-978 470311-980 470311-982 470311-984 470311-986 470311-988 470311-990 470311-992 470311-994 470311-996 470311-998 470312-000 470312-004 470312-006 470359-144 470359-152
3B Scientific® Birthing Simulators
Models Human Biology Models
The 3B Scientific® Birthing Simulator has been developed for the skill training in normal deliveries, in complicated deliveries and in obstetric emergencies.
  • Enhance the training of delivery skills, following of protocols and reaction in emergency situations
Obstetric simulation has proven successful to enhance the training of delivery skills, following of protocols and reaction in emergency situation.
Assessment and manipulation of fetal positions:
Birthing complications are in general less likely when an abnormal fetal position or presentation canMore Product Information