3B Scientific® Foot Structure Models
Supplier: American 3B Scientific
1000354 1000355 1000356 1024387 1024388 1024389
470029-530EA 257.4 CAD
470029-530 470118-882 470118-884 470359-084 470359-086 470359-088
3B Scientific® Foot Structure Models
Models Human Biology Models
Compare The Anatomy Of Foot Arch Problems
  • Includes 3B Smart Anatomy
Each of these models show the bone, muscle, ligament, and nerve structure of the foot. When compared differences in bone positon and muscular thickness can be seen. Presented as extremes, the Hollow foot and Flat foot versions magnify these differences.
Accessories information: For information about 3B Smart Anatomy, please see the document attached on this page.