Chicken Embryology (Gallus domesticus) - English Slides
Supplier: American 3B Scientific
1003986
470357-036EA 592.5 CAD
470357-036
Chicken Embryology (Gallus domesticus) - English Slides
Slides Prepared Slides
Ten microscope slides.
1(f). Chicken, 24 hour, t.s. with neural groove, notochord, germ layers
2(f). Chicken, 36 hour, t.s. with neural tube
3(g). Chicken, 48 hour, l.s. with differentiation of mesoderm and ectoderm
4(f). Chicken, 3 days, t.s. through body showing amnion and serosa
5(f). Chicken, 3 days, t.s. of head with primordium of brain, eyes and heart
6(g). Chicken, 3 to 4 days, horizontal section of entire specimen shows primordia of organs
7(f). Chicken, 4 to 5 days, t.s. region of head with brain, gill arches
8(f). Chicken, 4 to 5 days, t.s. region of heart
9(g). Chicken, 8 days,More Product Information