Magnaprobe with Magnetic Globe
Supplier: United Scientific Supplies
MGNGLBPRBVWR
470352-176EA 72.16 CAD
470352-176
Magnaprobe with Magnetic Globe
Globes
Visually demonstrate the magnetic field near the Earth.
  • Smooth action magnaprobe
  • Detailed globe with countries
  • Includes activity guide
The earth acts like it has a huge bar magnet located at its center. This set contains a globe that is 110 mm wide with a magnet contained along its axis. The magnaprobe contains a small magnetthat is allowed to rotate on two axes. This set illustrates the path of the magnetic field along the surface of a globe. The closer to the poles, the steeper the angle of the magnetic field. This is useful in demonstrating theMore Product Information