Ward's® Luster Collection
Supplier: Avantor
470315-518KT 58.3 CAD
470315-518
Ward's® Luster Collection
Specimens Mineral Specimens Mineral Collections
Explore the differences of how light interacts with the surface of a mineral.
The minerals in this collection are each perfect examples of one type of luster. The set is housed in a compartmented collection box, and an identification list is included.
Contents: 12 numbered samples, 1×1 to 1×2"
Metallic Luster - Pyrite, Chalcopyrite
Vitreous Luster - Quartz
Adamantine Luster - Sphalerite
Resinous Luster - Sulfur
Greasy Luster - Nepheline
Pearly Luster - Muscovite
Silky Luster - Satin spar
Splendent Luster - Micaceous hematite
Shining Luster - Calcite
Waxy Luster - Chalcedony
Submetallic Luster -More Product Information