Tube Holder S Student Series
Supplier: American 3B Scientific
1014525
470162-704EA 1473.5 CAD
470162-704
Tube Holder S Student Series
Educational Materials Physics Educational Materials Modern Physics Learning Activities
Easy and safe way to hold and power your tubes.
  • Easy and safe operation
  • Cathode protection switch
Tube holder to support all S series (student) electron tubes for easy and safe operation. The five-pin sockets for the tube are concealed inside the tube holder. A cathode protection switch is integrated into the tube holder, to protect the heated cathode from excessive voltage. The base plate has a slot for attaching the students’ Helmholtz coil pair. Terminals: 4 mm safety sockets.