Baseline® Three Point Descriminator
Supplier: American 3B Scientific
1017422
470119-640EA 94.7 CAD
470119-640
Baseline® Three Point Descriminator
Educational Materials Biology Educational Materials Physiology Learning Activities
Accurately test two point sensitivity.
  • Sliding point measured in mm
  • Switch between one and two point touch easily
Evaluate cutaneous sensitivity and touch threshold. Plastic points minimize influence of temperature. Two point discriminator has third point that permits alternating between single and double point stimulation without changing setting, measures up to 14 cm discrimination.
Order Now
SPECIFICATIONS
Environmentally Preferable