Comprehensive Mineral and Physical Properties Collection
Supplier: SCOTT RESOURCES 2, LLC SE
2340
470112-328KT 603.5 CAD
470112-328
Comprehensive Mineral and Physical Properties Collection
Specimens Mineral Specimens Mineral Collections
Teach students how to identify minerals with this collection.
  • Covers multiple mineral properties
  • Specimen lists included
Teach your students how to identify minerals by their physical properties with this extensive 126-sample collection. Each 1-1/2" x 1-1/2" specimen was chosen to show the following characteristics: cleavage and fracture, luster, hardness and tenacity, color, streak crystalline aggregates, fluorescence (short and long wave), specific gravity, other properties such as optical, thermal and magnetic. Specimens are packed in compartmentedMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable