Urinary Test Strips
Supplier: LW SCIENTIFIC SE
URS-01PR-BL77
470019-032EA 30.95 CAD
470019-032
Urinary Test Strips
Clinical Diagnostic Supplies Clinical Reagents and Kits
Includes Color Key for Easy Reading of Results
Simply place a drop of urine on the strip’s indicator spot and wait one minute for the color change. These strips are clinically tested for accuracy and have a shelf life of two years if kept in the resealable dry containers. They can also be used with WARD’S Artificial Urine. Vials of 100.