Coventry™ Pillow Tipped Swabs, ITW Chemtronics®
Supplier: ITW Chemtronics
52121 51121
15145-603CS 1149.77 CAD
15145-603 15145-605
Coventry™ Pillow Tipped Swabs, ITW Chemtronics®
Swabs Swabs for Cleaning
Pillow-tip™ swabs consist of polyester knit fabric which is held in place using a shrink tube
  • Engineered for high solvent retention with the cleanliness of ISO Class 4 processed polyester
  • Seamless design is lint-free
  • High-strength, adhesive-free swab, durable head construction
  • Excellent solvent compatibility
  • Available Aqua-Prime™ processed
Suitable for cleaning high end disk drive components, Aerospace Industry and Micro Mechanical cleaning, and to validate cleanliness of manufacturing surfaces for the Pharmaceutical and Biotechnical Industry.