Anti-IL7R Rat Monoclonal Antibody [clone: A7R34]

Supplier: Prosci
76-906
10765-008EA 605.35 CAD
10765-008
Anti-IL7R Rat Monoclonal Antibody [clone: A7R34]
Antibodies
The A7R34 monoclonal antibody specifically reacts with the mouse CD127, the alpha subunit of the IL-7 receptor, expressed by immature B lymphocytes in the bone marrow, CD4-/CD8-, CD4+, and CD8+ thymocytes, and by mature T lymphocytes at low levels. Mature T cells express CD117 at low levels in the periphery.The A7R34 antibody prevents the interaction between IL-7 and its receptor and the binding of mAb SB/199, which also recognizes mouse CD127.

Type: Primary
Antigen: Il7r
Clonality: Monoclonal
Clone: A7R34
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: Rat IgG2a, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR