Anti-CD3D Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: G4.18]

Supplier: Prosci
76-238
10764-228EA 1041.76 CAD
10764-228
Anti-CD3D Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: G4.18]
Antibodies
The G4.18 monoclonal antibody specifically reacts with rat CD3, a T cell lineage marker and associated with the T cell receptor (TCR). CD3 is involved in antigen recognition, cell activation, and signal transduction and is found on subsets of NK-T, peripheral T, dendritic epidermal T, and thymocyte cells. Immobilized G4.18 antibody activates T cells in vitro and in vivo treatments of the G4.18 antibody prevents cardiac and skin allograft rejection.

Type: Primary
Antigen: Cd3d
Clonality: Monoclonal
Clone: G4.18
Conjugation: FITC
Epitope:
Host: Mouse
Isotype: Mouse IgG3, kappa
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR