Anti-EMR1 Rat Monoclonal Antibody (violetFluor® 450) [clone: BM8.1]
Supplier: Prosci
76-056
10764-112EA 666.13 CAD
10764-112
Anti-EMR1 Rat Monoclonal Antibody (violetFluor® 450) [clone: BM8.1]
Antibodies
The BM8.1 monoclonal antibody specifically binds to the Mouse125 kDa F4/80 antigen, expressed by most mature macrophages. F4/80 is a transmembrane protein used as a marker of macrophages, although it is also expressed on Kupffer and Langerhans cells. The expression of F4/80 antigen is upregulated on bone marrow cells stimulated in vitro with the macrophage colony stimulating factor. The F4/80 antigen is a requirement for the induction of CD8 T cells-mediated peripheral tolerance.BG Violet 450 conjugate is an alternative to the Pacific Blue, eFluor 450, or BD Horizon V450 dyes. It is excitedMore Product Information