Anti-CMYA5 Rabbit Polyclonal Antibody
Supplier: Prosci
6403_S
10751-366EA 310.85 CAD
10751-366
Anti-CMYA5 Rabbit Polyclonal Antibody
Antibodies
SPRYD2 Antibody: SPRYD2, also known as Myospryn, was originally identified as the muscle-specific partner of dysbindin and as a Mef-2 target gene. It is a large scaffolding protein localized to the Z-disc/costamere region of striated muscle. SPRYD2 includes a noncanonical tripartite motif (TRIM-like) that lacks the RING domain but consists of a B-box coiled coil (BBC), fibronectin 3 (FN3) repeats, and SPRY domains. SPRYD2 interacts with desmin and calcineurin, and it has been suggested to play a role in the biogenesis of lysosome and negatively regulates slow-fiber-type transformation andMore Product Information