Anti-APOA5 Rabbit Polyclonal Antibody

Supplier: Prosci
6271_S
10751-298EA 310.85 CAD
10751-298
Anti-APOA5 Rabbit Polyclonal Antibody
Antibodies
ApoA5 Antibody: Apolipoprotein A4 (also known as ApoA-IV) is a plasma protein that is O-linked glycoprotein after proteolytic processing. ApoA4 is a major constituent of HDL and it also can be found on the surface of newly synthesized chylomicrons. The physiological function of ApoA4 is not completely identified so far, but several evidences from vitro studies indicate that this apolipoprotein involves in several steps of the reverse cholesterol transport pathway, which transports cholesterol from peripheral cells back to the liver.

Type: Primary
Antigen: APOA5
Clonality: Polyclonal
Clone:
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR