Anti-LRRTM4 Rabbit Polyclonal Antibody
Supplier: Prosci
6173_S
10751-226EA 310.85 CAD
10751-226
Anti-LRRTM4 Rabbit Polyclonal Antibody
Antibodies
LRRTM4 Antibody: The Leucine-rich repeat transmembrane neuronal proteins (LRRTMs) are differentially expressed in the nervous system and were recently found to instruct presynaptic and mediate postsynaptic glutamatergic differentiation, with LRRTM1 and LRRTM2 most potent at inducing presynaptic differentiation. Little is known about the function of LRRTM4; in mouse, it is expressed in the limb mesenchyme, neural tube, caudal mesoderm and in three distinct regions of the head. Later expression occurs in a subset of the developing sclerotome. Its similarity to other LRRTM family membersMore Product Information