Anti-SLAIN1 Rabbit Polyclonal Antibody

Supplier: Prosci
5467_S
10750-712EA 301.81 CAD
10750-712
Anti-SLAIN1 Rabbit Polyclonal Antibody
Antibodies
SLAIN1 Antibody: SLAIN1 is a novel stem cell protein that was identified by transciptional profiling of mouse and human embryonic stem cells (ESCs) and is expressed at the stem cell and epiblast stages. Unlike its more widely expressed homolog SLAIN2, SLAIN1 was found to be expressed in the epiblast, nervous system, tailbud and somites of the developing mouse embryo later experiments suggested that SLAIN1 may play a role in the development of the nervous system as well as in the morphogenesis of several embryonic structures.

Type: Primary
Antigen: SLAIN1
Clonality: Polyclonal
Clone:
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR