Anti-ANO1 Rabbit Polyclonal Antibody
Supplier: Prosci
5419_S
10750-674EA 301.81 CAD
10750-674
Anti-ANO1 Rabbit Polyclonal Antibody
Antibodies
TMEM16A Antibody: Calcium-activated chloride channels (CaCC) are present in many cell types and mediate physiological functions such as epithelial secretion, sensory signal transduction, and smooth muscle contraction. Subunits of these CaCC's include the transmembrane proteins TMEM16A and TMEM16B. TMEM16A is expressed in epithelial cells of the kidney and lung, pancreas, and sensory neurons and its mRNA is seen in foregut, airway epithelia, and tracheal smooth muscle; mice lacking TMEM16A fail to survive past ten days and show aerophagia and little weight gain. TMEM16A is also overexpressedMore Product Information