Anti-TICAM2 Rabbit Polyclonal Antibody
Supplier: Prosci
3206_S
10749-568EA 301.81 CAD
10749-568
Anti-TICAM2 Rabbit Polyclonal Antibody
Antibodies
TIRP Antibody: TIRP is a member of the Toll/interleukin-1 receptor (TIR) family, a group of proteins that include the Toll-like receptors (TLRs). TLRs are signaling molecules that recognize different pathogen-associated molecular patterns (PAMPs) and serve as an important link between the innate and adaptive immune responses. TIRP, along with other molecules such as TRIF, MAL, and MyD88, serves as an adaptor protein that allows for the interaction and activation of the IL-1R-associated kinase (IRAK) family, the subsequent activation of TNF receptor associated factor (TRAF)-6, and ultimatelyMore Product Information