Anti-WNVgp1 Rabbit Polyclonal Antibody
Supplier: Prosci
3431_S
10748-420EA 301.81 CAD
10748-420
Anti-WNVgp1 Rabbit Polyclonal Antibody
Antibodies
West Nile Virus Matrix Antibody: West Nile Virus (WNV) is a member of the Flaviviridae, a plus-stranded virus family that includes St. Louis encephalitis virus, yellow fever virus, and Dengue virus. WNV was initially isolated in 1937 in the West Nile region of Uganda and has become prevalent in Africa, Asia, and Europe. It has rapidly spread across the United States with cases being observed in every continental state. Virus particles consist of a dense core made up of the core/capsid protein encapsulating the RNA genome surrounded by a membrane envelope embedded with envelope and matrixMore Product Information