Anti-IQCG Rabbit Polyclonal Antibody
Supplier: Bioss
BS-9022R
10671-464EA 997.44 CAD
10671-464
Anti-IQCG Rabbit Polyclonal Antibody
Antibodies
IQCG is a 443 amino acid protein containing one IQ domain. Widely distributed in nature, the IQ domain forms an amphiphilic seven-turn α-helix capable of binding calmodulin in a Ca2+-independent manner. The level of intracellular calcium is tightly regulated in all eukaryotic cells. A modest increase in this level can result in a myriad of physiological responses, most of which are mediated by calmodulin (CaM), the universal calcium sensor. In acute T-lymphoid/myeloid leukemia, IQCG forms a complex with Nup98, an O-linked glycoprotein and a component of the nuclear pore complex. NUP98-IQCGMore Product Information