Anti-ZDHHC18 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: Bioss
BS-8641R-HRP
10671-042EA 1270.19 CAD
10671-042
Anti-ZDHHC18 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. ZDHHC1 (zinc finger, DHHC-type containing 1), also known as ZNF377 (zinc finger protein 377) or C16orf1, is a 485 amino acid multi-pass membrane protein that contains one DHHC-type zinc finger. Expressed in fetal heart, lung and kidney and also present in adult lung and pancreas, ZDHHC1 functions as a palmitoyltransferase that catalyzes the conversion of palmitoyl-CoA and protein-cysteine to S-palmitoyl protein and CoA. LikeMore Product Information