Anti-TRPM6 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-9048R-CY7
10666-176EA 1269.81 CAD
10666-176
Anti-TRPM6 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
TRPM6 is an essential ion channel and serine/threonine-protein kinase, and is crucial for magnesium homeostasis. TRPM6 also has an important role in epithelial magnesium transport and in the active magnesium absorption in the gut and kidney. The various isoforms of the type M6-kinase lack the ion channel region.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TRPM6
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR