Anti-MEPE Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: Bioss
BS-8689R-HRP
10664-610EA 1270.19 CAD
10664-610
Anti-MEPE Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
MEPE is a 525 amino acid extracellular matrix protein. Expressed in osteocytes and brain, MEPE is a regulator of bone metabolism that is thought to mediate mineralization and demineralization within the osteocyte microenvironment. MEPE contains an RGD cell-attachment motif and shares molecular similarities with several dentin-bone extracellular matrix RGD-containing phosphoglycoproteins, including OPN (osteopontin) and DSP (dentin sialophosphoprotein). Via its ability to control bone mineralization, MEPE is associated with various developmental events such as skeletogenesis, boneMore Product Information