Anti-TAS1R1 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-8612R-CY7
10663-964EA 1270.21 CAD
10663-964
Anti-TAS1R1 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Putative taste receptor. TAS1R1/TAS1R3 responds to the umami taste stimulus (the taste of monosodium glutamate). Sequence differences within and between species can significantly influence the selectivity and specificity of taste responses.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TAS1R1
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR