Anti-ARG1 Rabbit Polyclonal Antibody (Cy5.5®)
Supplier: Bioss
BS-8585R-CY5.5
10663-528EA 1270.21 CAD
10663-528
Anti-ARG1 Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
Arginase I which is expressed almost exclusively in the liver, catalyzes the conversion of arginine to ornithine and urea . The human arginase I gene, which maps to chromosome 6q23, encodes a 322 amino acid protein. Arginase I exists as a homotrimeric protein and contains a binuclear manganese cluster. Arginase II catalyzes the same reaction as arginase I, but differs in its tissue specificity and subcellular location. Specifically, arginase II localizes to the mitochondria. Arginase II is expressed in non-hepatic tissues, with the highest levels of expression in the kidneys, but, unlikeMore Product Information