Anti-CLEC4E Rabbit Polyclonal Antibody
Supplier: Bioss
BS-8541R
10662-722EA 997.45 CAD
10662-722
Anti-CLEC4E Rabbit Polyclonal Antibody
Antibodies
C-type lectin that functions as cell-surface receptor for a wide variety of ligands such as damaged cells, fungi and mycobacteria. Plays a role in the recognition of pathogenic fungi, such as Candida albicans. The detection of mycobacteria is via trehalose 6,6'-dimycolate (TDM), a cell wall glycolipid. Specifically recognizes alpha-mannose residues on pathogenic fungi of the genus Malassezia. Recognizes also SAP130, a nuclear protein, that is released by dead or dying cells. Transduces signals through an ITAM-containing adapter protein, Fc receptor gamma chain /FCER1G. Induces secretion ofMore Product Information