Anti-SIRT5 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss

BS-9456R-CY5
10495-390EA 1270.21 CAD
10495-390
Anti-SIRT5 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
NAD-dependent lysine demalonylase and desuccinylase that specifically removes malonyl and succinyl groups on target proteins. Activates CPS1 and contributes to the regulation of blood ammonia levels during prolonged fasting: acts by mediating desuccinylation of CPS1, thereby increasing CPS1 activity in response to elevated NAD levels during fasting. Activates SOD1 by mediating its desuccinylation, leading to reduced reactive oxygen species. Has weak NAD-dependent protein deacetylase activity; however this activity may not be physiologically relevant in vivo. Can deacetylate cytochrome c (More Product Information
Order Now

SPECIFICATIONS

Antigen symbol SIRT5
Environmentally Preferable
Antigen name Sirtuin 5
Antigen synonyms SIR2L5|NAD-dependent protein deacylase sirtuin-5|Regulatory protein SIR2 homolog 5|mitochondrial
Antibody type Primary
Clonality Polyclonal
Conjugation Cy5®
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 23408
Isotype IgG
ImmunoChemistry Yes
Immunogen 120-170/310
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at –20 °C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR