Anti-GPR71 Rabbit Polyclonal Antibody (Cy3®)
Supplier: Bioss
BS-9599R-CY3
10492-074EA 1270.19 CAD
10492-074
Anti-GPR71 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
The sense of taste provides animals with valuable information about the quality and nutritional value of food. There are four widely accepted categories of taste perception, sweet, bitter, salty, and sour. A controversial fifth taste, known as umami or monosodium glutamate (MSG), has also been described. A family of G protein coupled receptors are involved in taste perception, and includes T1R, which is involved in sweet and umami taste perception, and T2R, which is involved in bitter taste perception. The T1R family consists of three members, T1R1, T1R2, and T1R3 (1-4). These proteins formMore Product Information