Anti-CNOT2 Rabbit Polyclonal Antibody (Cy3®)
Supplier: Bioss
BS-9383R-CY3
10489-426EA 1270.19 CAD
10489-426
Anti-CNOT2 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
CNOT2 (CCR4-NOT transcription complex subunit 2) is a ubiquitous protein encoded by the human gene CNOT2. CNOT2 belongs to the CNOT2/3/5 family and is part of the CCR4-NOT complex. The CCR4-NOT complex is an evolutionarily conserved, multi-component complex known to be involved in transcription as well as mRNA degradation. Various subunits (e.g. CNOT1, CNOT2) are involved in influencing nuclear hormone receptor activities. The CCR4-NOT complex is also involved in the regulation of Histone H3 lysine 4 methylation through a ubiquitin-dependent pathway that likely involves the proteasome.More Product Information