Anti-MARCH9 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-9343R-CY7
10489-122EA 1270.19 CAD
10489-122
Anti-MARCH9 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Ubiquitination is an important mechanism through which three classes of enzymes act in concert to target short-lived or abnormal proteins for destruction. The three classes of enzymes involved in ubiquitination are the ubiquitin-activating enzymes (E1s), the ubiquitin-conjugating enzymes (E2s) and the ubiquitin-protein ligases (E3s). MARCH9 (membrane-associated ring finger (C3HC4) 9), also known as RNF179, is a 346 amino acid multi-pass membrane protein that localizes to the golgi apparatus and contains one RING-CH-type zinc finger. Expressed ubiquitously, MARCH9 exists as a homodimer andMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol MARCH9
Antigen name Membrane-associated ring Finger (C3HC4)9, E3 ubiquitin protein ligase
Environmentally Preferable
Antigen synonyms Membrane associated RING finger protein 9|MARCH IX|membrane associated ring finger C3HC4 9|RING finger protein 179|Membrane associated RING CH protein IX|MARH9_HUMAN.|RNF179|E3 ubiquitin protein ligase MARCH9
Antibody type Primary
Clonality Polyclonal
Conjugation Cy7®
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 92979
Isotype IgG
ImmunoChemistry Yes
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at –20 °C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR