Anti-F11 Rabbit Polyclonal Antibody (Cy3®)
Supplier: Bioss
BS-9502R-CY3
10488-726EA 1270.21 CAD
10488-726
Anti-F11 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
This gene encodes coagulation factor XI of the blood coagulation cascade. This protein is present in plasma as a zymogen, which is a unique plasma coagulation enzyme because it exists as a homodimer consisting of two identical polypeptide chains linked by disulfide bonds. During activation of the plasma factor XI, an internal peptide bond is cleaved by factor XIIa (or XII) in each of the two chains, resulting in activated factor XIa, a serine protease composed of two heavy and two light chains held together by disulfide bonds. This activated plasma factor XI triggers the middle phase of theMore Product Information