Anti-LRRC2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Bioss

BS-8360R-HRP
10482-720EA 1270.19 CAD
10482-720
Anti-LRRC2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
The leucine-rich repeat-containing protein 2 (LRRC2) is a 371 amino acid protein that contains 9 LRR repeats. The gene encoding LRRC2 maps to chromosome 3, which encodes over 1,100 genes. Particular regions of the chromosome 3 short arm are deleted in many types of cancer cells. Marfan Syndrome, porphyria, von Hippel-Lindau syndrome, osteogenesis imperfecta and Charcot-Marie-Tooth Disease are a few of the numerous genetic diseases associated with chromosome 3.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: LRRC2
Clonality: Polyclonal
Clone:
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR