Anti-FRMPD1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Bioss

BS-8233R-FITC
10479-878EA 1270.19 CAD
10479-878
Anti-FRMPD1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
FRMPD1 has been identified as a regulatory binding partner of activator of G-protein signaling 3 (AGS3).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FRMD2
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR