Anti-CCDC158 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Bioss

BS-8122R-FITC
10477-344EA 1270.19 CAD
10477-344
Anti-CCDC158 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
CCDC158

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CCDC158
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

SPECIFICATIONS

Antigen symbol CCDC158
Environmentally Preferable
Antigen name coiled-coil Domain Containing 158
Antibody type Primary
Clonality Polyclonal
Conjugation FITC (Fluorescein Isothiocyanate)
Reactivity Human,Rat,Mouse
Host Rabbit
Isotype IgG
ImmunoChemistry Yes
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at –20 °C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR