Anti-ASNA1 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-7972R-CY7
10473-988EA 1270.19 CAD
10473-988
Anti-ASNA1 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
ASNA1 is the human homolog of the E.coli arsA gene which is an ATPase, and is the catalytic component of a multisubunit oxyanion pump responsible for resistance to arsenicals and antimonials.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ASNA1
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR