Anti-AKR1A1 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-7953R
10473-592EA 997.44 CAD
10473-592
Anti-AKR1A1 Rabbit Polyclonal Antibody
Antibodies
Catalyzes the NADPH-dependent reduction of a variety of aromatic and aliphatic aldehydes to their corresponding alcohols. Catalyzes the reduction of mevaldate to mevalonic acid and of glyceraldehyde to glycerol. Has broad substrate specificity. In vitro substrates include succinic semialdehyde, 4-nitrobenzaldehyde, 1,2-naphthoquinone, methylglyoxal, and D-glucuronic acid. Plays a role in the activation of procarcinogens, such as polycyclic aromatic hydrocarbon trans-dihydrodiols, and in the metabolism of various xenobiotics and drugs, including the anthracyclines doxorubicin (DOX) andMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol AKR1A1
Antigen name Aldo-keto reductase family 1, member A1
Environmentally Preferable
Antigen synonyms member A1|member A1 (Aldehydereductase)|AKR1A1 protein|isoform CRA_a|Aldehyde reductase|Aldo-keto reductase family 1
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 10327
Isotype IgG
Western blot Yes
ImmunoChemistry Yes
Immunogen 72-102/325
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at -20°C for 12 months.
Storage temperature Store at –20 °C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR