Anti-CDCA2 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-7786R-CY3
10469-734EA 1270.19 CAD
10469-734
Anti-CDCA2 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Regulator of chromosome structure during mitosis required for condensin-depleted chromosomes to retain their compact architecture through anaphase. Acts by mediating the recruitment of phopsphatase PP1-gamma subunit (PPP1CC) to chromatin at anaphase and into the following interphase. At anaphase onset, its association with chromatin targets a pool of PPP1CC to dephosphorylate substrates.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CDCA2
Clonality: Polyclonal
Clone: 0
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

SPECIFICATIONS

Antigen symbol CDCA2
Environmentally Preferable
Antigen name Cell Division Cycle associated 2
Antigen synonyms recruits PP1 onto mitotic chromatin at anaphase protein|Repo-Man|protein phosphatase 1 [gamma] (PPP1R3A) binding protein Repo-Man|MGC129907|MGC129906|Cell Division Cycle-associated protein 2|FLJ25804
Antibody type Primary
Clonality Polyclonal
Conjugation Cy3®
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 157313
Isotype IgG
ImmunoChemistry Yes
Immunogen 373-410/1023
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at –20 °C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR