Anti-NIR2 Rabbit Polyclonal Antibody (Cy5®)
Supplier: Bioss
BS-7763R-CY5
10469-048EA 1270.19 CAD
10469-048
Anti-NIR2 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
Regulates RHOA activity, and plays a role in cytoskeleton remodeling. Necessary for normal completion of cytokinesis. Plays a role in maintaining normal diacylglycerol levels in the Golgi apparatus. Binds phosphatidyl inositol phosphates (in vitro). May catalyze the transfer of phosphatidylinositol and phosphatidylcholine between membranes (By similarity). Necessary for maintaining the normal structure of the endoplasmic reticulum and the Golgi apparatus. Required for protein export from the endoplasmic reticulum and the Golgi. Binds calcium ions.
Recommended Dilutions: IF(IHC-P): 1:50-200More Product Information